Skip to Content
Merck
All Photos(1)

Key Documents

S7109

Sigma-Aldrich

Propidium Iodide Solution

Synonym(s):

3,8-diamino-5-[3-(diethylmethylammonio)propyl]-6-phenylphenanthridinium Diiodide

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
1.880,00 kr.

1.880,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
1.880,00 kr.

About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.41

1.880,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

manufacturer/tradename

Chemicon®

Quality Level

technique(s)

activity assay: suitable (apoptosis)

detection method

fluorometric

shipped in

dry ice

General description

{3,8-diamino-5-[3-(diethylmethylammonio)propyl]-6-phenylphenanthridinium diiodide}.

250 μg/ml Propidium Iodide (PI) in PBS.

Application

Propidium Iodide is a nuclear fluorescent stain used for counterstaining paraffin-embedded, formalin-fixed tissue, to discriminate between live vs. dead cells in flow cytometry, and staining metaphase chromosomes or interphase nuclei that have been hybridized with probes detected with Fluorescein, or equivalent fluors.



Materials Provided

Each vial contains 1 ml of Propidium Iodide Solution



Warnings and Precautions

Propidium Iodide is a potential carcinogen. Use extreme caution. Wear gloves and a lab coat when handling. Follow your institution′s guidelines for hazardous waste disposal. Keep tightly capped and limit exposure to light.



Related Products

Antifade Solution S7114

DAPI/Antifade Solution S7113

Propidium Iodide/Antifade Solution S7112

Storage and Stability

Recommended Storage

-15ºC to -25ºC

Protect from light

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service