Skip to Content
Merck
All Photos(1)

Key Documents

S4030-M

Millipore

Dextran Sulfate Solution

50% (w/v) liquid

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
282.600,00 kr.

282.600,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
100 ML
282.600,00 kr.

About This Item

UNSPSC Code:
41116012
eCl@ss:
32160405
NACRES:
NA.31

282.600,00 kr.


Please contact Customer Service for Availability

Product Name

Dextran Sulfate 50% Solution,

packaging

pkg of 100 mL

Quality Level

manufacturer/tradename

Chemicon®

technique(s)

hybridization: suitable (nucleic acid)

General description

Dextran Sulfate (MW >500,000) is used as an additive in probe hybridization solutions. Its presence accelerates the rate of hybridization by 10 fold. The addition of Dextran Sulfate favors the formation of probe networks, resulting in greater sensitivity during membrane-bound nucleic acid analysis. The product is supplied as a 50% (w/v) solution.

Application

Dextran Sulfate - 50% Solution

To prepare a solution containing 10% Dextran Sulfate, add 8 ml of hybridization ingredients to 2 ml of 50% Dextran Sulfate Solution.



See the "HYBRISOL® Nomograph for Dextran Sulfate" link located under the "Protocols" tab at catalog number S4030 product specific page on www.chemicon.com for determining the proper concentration of dextran sulfate needed for various hybridization condition guidelines.
Research Category
All

Physical form

A 50% dextran sulfate solution in water filtered through a 20 micron membrane and autoclaved prior to bottling in clean, sterilized bottles containing no preservatives. Liquid solution is dark brown from autoclaving.

Storage and Stability

Store at room temperature tightly sealed when not in use. Solution can be stored refrigerated however it should be warmed to at least room temperature prior to use dilution for best results.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
HYBRISOL is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service