RDYTUBE08
Set of 8 Tubes
For connection of adapters to GL45 cap. To be used in ReadyStream®workflow.
Synonym(s):
Silicone tubes
Select a Size
873,00 kr.
Estimated to ship on28 March 2025Details
Select a Size
About This Item
873,00 kr.
Estimated to ship on28 March 2025Details
Recommended Products
material
silicone
Sterilization Compatibility
autoclavable compatible
packaging
pkg of 8 tubes
application(s)
food and beverages
General description
Legal Information
related product
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service