Skip to Content
Merck
All Photos(1)

Key Documents

PYR0CNT01

Milli-Q

Pyrogard® D Point-of-Use Connector

For use with Pyrogard® D ultrafiltration cartridge

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
872,00 kr.

872,00 kr.


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 EA
872,00 kr.

About This Item

UNSPSC Code:
41104214
eCl@ss:
33050190
NACRES:
JA.13

872,00 kr.


Please contact Customer Service for Availability

Request more information

packaging

pkg of 1 unit

technique(s)

sterile filtration: suitable (cell culture | mammalian)

pore size

0.45 μm pore size (CRW5 type)

fitting

1/4 in. inlet connection
1/4 in. outlet connection

compatibility

for use with Milli-Q® IQ Element
for use with Milli-Q® Academic
for use with Milli-Q® Biocel
for use with Milli-Q® Gradient
for use with Milli-Q® Integral
for use with Milli-Q® Synthesis

shipped in

ambient

General description

Pyrogard® D Point-of-Use Connector is a Water Purification Accessory, used to connect Pyrogard® D ultrafiltration cartridge with Milli-Q® Academic / Biocel / Element / Gradient / Synthesis laboratory water system.

Application

Pyrogard® D Point-of-Use Connector is used for leak free fitting of Pyrogard® D ultrafiltration cartridge.

Features and Benefits

To be used only with Pyrogard® D ultrafiltration cartridge for Milli-Q® Academic / Biocel / Element / Gradient / Synthesis laboratory water system. Available with O-ring.

Legal Information

Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
PYROGARD is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service