Skip to Content
Merck
All Photos(2)

Documents

PR0GTXLCS2

Milli-Q

Progard® XL Pretreatment Pack

For optimal protection of reverse osmosis (RO) membranes from hard water

Synonym(s):

Progard® XL-S-C Pretreatment Pack

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104200
NACRES:
NB.85

material

carbon cartridge (activated )
cartridge

Quality Level

packaging

pkg of 2 units

manufacturer/tradename

Progard®

parameter

2-40 °C temp. range
4.2 L/min flow rate
6 bar max. pressure

H × W × D

20.8 in. × 6.1 in. × 3.1 in.

fitting

11 mm female inlet and outlet (O-rings)

compatibility

for use with AFS®
for use with Elix® Large
for use with Milli-Q® CLX
for use with Milli-Q® HR
for use with Milli-Q® HX
for use with RiOs Large

Looking for similar products? Visit Product Comparison Guide

General description

Polyphosphate for anti-scaling protection.
Silver-impregnated activated carbon for free chlorine reduction and bacteriostatic protection.
The Progard® XL-S-C pretreatment pack pretreats feed water by removing large particles and free chlorine from tap water. It protects the reverse osmosis (RO) membrane against mineral scaling, organic fouling, and chlorine oxidation.
The pretreatment pack includes prefiltration to remove particles, activated carbon to remove chlorine, and an anti-scaling agent to minimize the risk of scale formation. The Autoclean pack option includes a built-in chlorine cleaning tablet that will automatically clean the RO membrane when this new pack is installed.
For use with larger RiOs, Elix®, Milli-Q® HR, HX, CLX and AFS® E water purification systems.

Application

The packs protect and maintain good performance of RO membranes in water purification systems.

Features and Benefits

  • Optimal protection of reverse osmosis (RO) membrane
  • Ensures system lifetime and performance are optimized
  • Depth-filtration removes particles and colloids.
  • Polyphosphate protects from scaling.
  • Silver-impregnated activated carbon filter reduces free chlorine and provides bacteriostatic protection.
  • EfferSanEffervescent (formerly ROProtect CR) tablets provide automatic RO maintenance
  • e-Sure tag RFID technology for instant visibility on pack parameters, automatic pack parameters archiving and full traceability.
  • Replace every 6 months for optimal results

Legal Information

AFS is a registered trademark of Merck KGaA, Darmstadt, Germany
ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
PROGARD is a registered trademark of Merck KGaA, Darmstadt, Germany
RIOS is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service