Skip to Content
Merck
All Photos(1)

Documents

PBQK50205

Millipore

Biomax® Ultrafiltration Membrane

Ultrafiltration, high flux,low protein binding, chemical compatibility

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104903
eCl@ss:
32161003
NACRES:
NB.24

material

polyethersulfone membrane

Quality Level

filtration area

20 cm2

membrane L × W

20 cm × 20 cm

matrix

Biomax®

pore size

50 kDa NMWCO

application(s)

industrial qc (particle separation)

shipped in

ambient

General description

Device Configuration: Ultrafiltration Membrane
Filter Code: PBQK

Application

Ultrafiltration, high flux,low protein binding, chemical compatibility

Legal Information

BIOMAX is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Free eBook outlines plasmid DNA manufacturing process and downstream purification challenges for various applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service