P3B050A10
Pellicon® 3 Cassette with Biomax® Membrane
50kDa, A Screen, 1.14 m2
Synonym(s):
Pellicon® 3 Cassette withBiomax 50 kDa Membrane, A screen, 1.14 m2
About This Item
Recommended Products
material
TPE seal
polyethersulfone
polypropylene screen
stainless steel cap (for end; polypropylene-encapsulated)
Quality Level
feature
holdup volume 134 mL (in feed channel)
manufacturer/tradename
Pellicon®
parameter
30 psi max. reverse press. exposure at 25 °C (for 24 hours)
4-6 L/min-m2 recirculation rate
40 psi max. transmembrane pressure at 4-50 °C
6.9 bar max. inlet pressure (100 psi)
80 psi max. transmembrane pressure at 4-40 °C
technique(s)
analytical sample preparation: suitable
buffer exchange: suitable
protein purification: suitable (concentration)
H
3.8 cm (1.51 in.)
L
20.6 cm (8.1 in.)
W
17.8 cm (7 in.)
membrane area
1.14 m2
gravimetric extractables
≤1500 mg/m2
matrix
Biomax® (Membrane)
pore size
50 kDa NMWCO
pH
1-14
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
The Pellicon® 3 Cassette with Biomax® Membrane is perfect for the filtration of monoclonal antibodies and solutions containing albumin, hormones, vaccines and growth factors.
This P3B050A10 Pellicon® 3 ultrafiltration device features a pore size of 50 kDa NMWCO, a Biomax® 50 membrane, and dimensions measuring 3.8 cm (H) x 17.8 cm (W) x 20.6 cm (L).
Features and Benefits
- Rugged & reliable – ideally suited for today′s more demanding filtration processes
- High performance – produces excellent resolution, yields and back-pressure resistance
- Optimum product recovery – thanks to proven membrane technology
- Void-free technology – leading-edge membranes resolve virtually any separation challenge
- Defect-free, superior adhesion – Ultracel®uses composite membranes cast on a microporous substrate
- Automated manufacturing – delivers unbeatable performance, consistency and reliability
- Easy installation & care – simple to install and maintain
Other Notes
Analysis Note
Legal Information
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service