P2B010V01
Pellicon® 2 Mini Ultrafiltration Biomax® Module
filtration area 0.1 m2, NMWCO 10 kDa
Synonym(s):
Pellicon® 2 Mini Ultrafiltration Module Biomax-10 V 0.1 m2
About This Item
Recommended Products
material
polyethersulfone
polypropylene screen
polyurethane adhesive
Quality Level
manufacturer/tradename
Pellicon®
parameter
0.33 bar max. transmembrane pressure (5 psig) (reverse)
5-35 L/min recirculation rate at 0.4 bar (6 psi)
7 bar max. transmembrane pressure (100 psig) at 30 °C (forward)
technique(s)
analytical sample preparation: suitable
protein purification: suitable
H
2.16 cm (0.85 in.)
L
21 cm (8.3 in.)
W
5.6 cm (2.2 in.)
filtration area
0.1 m2
membrane nominal area
0.1 m2
gravimetric extractables
≤1500 mg/m2
matrix
Biomax®
pore size
10 kDa NMWCO
pH
1-14
Looking for similar products? Visit Product Comparison Guide
General description
Millipore′s BIOMAX 10 polyethersulfone membrane has void-free structures that guard against leakage of solutes through microdefects normally associated with voids beneath the thin skins of conventional UF membranes. These void-free membranes are more permeable, resulting in high-flux with equivalent or superior product retention. These void-free membranes provide the advantages of fast, high yield processing and smaller systems.
The Pellicon® 2 Mini Ultrafiltration BIOMAX Module feature a filtration area of 0.1 m2 with a pore size of 10 kDa NMWCO.
Application
- Protein solutions
- Polysaccharide solutions
- Virus suspensions
Harvest, washing or clarification of:
- Cell cultures and lysates
- Colloidal suspensions
- Viral cultures
Features and Benefits
- Type V screen (open channel)
- Leading-edge void- free membranes to match virtually any separation challenge
- Short flow path for higher flux and higher resolution separation capability
- Choice of flow channel configuration providing process optimization capability
- Predictable, fast, scale-up
- True linear scalability from laboratory size modules to industrial assemblies for processing thousands of liters
- Brings higher resolution, improved yields and superior back-pressure resistance
- Ultracel membranes are composite membranes cast on a microporous substrate for defect-free membranes with superior
Other Notes
Analysis Note
Legal Information
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service