NU050/31-221
NovAseptic® Shut off (1/2") Valve Body 90 °
Valve Body - ASTM® A269/270, Electropolished Butt end for welding
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
23151800
Quality Level
parameter
-1-10 bar max. pressure (-14.5-145 psig) at 25 °C (atmospheric pressure)
-80-200 °C max. temp. (-112-392 °F)
General description
NovAseptic® Valves are engineered for optimal performance, reliability, durability and ease of maintenance. NovAseptic® Valves comply with cGMP Design Qualification
criteria for aseptic processing. This is why we choose to call it ‘‘Aseptic by Design.”
criteria for aseptic processing. This is why we choose to call it ‘‘Aseptic by Design.”
Specificity
Net volume (Valve cavity, diaphragm applied): 4mLNet weight (Approximate): Weight kg (lb): 0.1(0.22)Surface roughness: Internal surface Ra ≤ 0.38 μm (15 μin) External surface Ra ≤ 1.6 μm (63 μin)
Application
NovAseptic® Valves are specifically designed for aseptic applications and comply with the most stringent cleanability and sterilizability requirements. The focus on aseptic design is a significant feature for all valves. The diaphragm seal system on the NovAseptic® Valve produces positive sealing with no cavities. The seal remains static regardless of the position of the diaphragm. The valve is totally drainable and cleanable. 90° or 180° shut-off valves give engineers full flexibility with full drainability and low pressure drop.
Components
All open ends on the valve body are protected with plastic covers on delivery and packaged in a closed box.
Legal Information
ASTM is a registered trademark of American Society for Testing and Materials
NOVASEPTIC is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service