NB-2ML-RU
Reagent For Research Use - Rare Specificities & AHG
BIOSCOT™ Anti-Lea from the clone GA2, Murine IgA, Blood Grouping Reagent for Research Use
Synonym(s):
Anti-Lea (clone GA2), Blood Grouping Reagent for Research Use
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116102
biological source
mouse
antibody form
unpurified
clone
GA2, monoclonal
form
liquid
packaging
pkg of 2 mL
concentration
3-12 mg/mL protein (Total Protein Range)
technique(s)
agglutination assay: suitable
pH
6-8
isotype
IgA
shipped in
ambient
storage temp.
2-8°C
General description
Contains 0.1% (w/v) Sodium Azide. Filtered through a 0.22μm terminal filter. Shelf Life: 36 months from date of filtration, For Non Clinical In Vitro research use only.
Application
For use by Tube technique
Vialled, Labeled
Legal Information
BIOSCOT is a trademark of Merck KGaA, Darmstadt, Germany
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service