Skip to Content
Merck
All Photos(2)

Key Documents

MXRPA4VP1

Millipore

Milliflex Rapid Detection and Enumeration System Validation Protocol

The Milliflex Rapid Detection/Enumeration System validation protocol makes validation faster, easier and ensures that your system′s performance remains compliant with its validated specifications.

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
17.150,00 kr.

17.150,00 kr.


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 EA
17.150,00 kr.

About This Item

UNSPSC Code:
41116500
eCl@ss:
25169090
NACRES:
NB.79

17.150,00 kr.


Please contact Customer Service for Availability

Request more information

Quality Level

manufacturer/tradename

Milliflex®

input

sample type pharmaceutical(s)

application(s)

industrial qc

shipped in

ambient

General description

Service Type: Validation Services

Application

The Milliflex Rapid Detection/Enumeration System validation protocol makes validation faster, easier and ensures that your system′s performance remains compliant with its validated specifications.

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service