MX0HC10FS27EU
Millistak+® HC Pod Depth Filter
X0HC, filtration area 1.1 m², Process Scale in EU format Bulk Packaging
Synonym(s):
Millistak+ Pod Depth Filter, X0HC media series, 1.1 m 2 surface area, 27/pack shipped on a European CP-2 standard sized pallet
Select a Size
183.933,00 kr.
Select a Size
About This Item
183.933,00 kr.
Recommended Products
Product Name
Millistak+® HC Pod Depth Filter, X0HC, cellulose, filtration area 1.1 m2, hydrophilic
material
cellulose
filter aid (inorganic)
glass fiber-reinforced polypropylene
Quality Level
description
Secondary clarification of bioreactor harvests, primarily for cell cultures
feature
hydrophilic
manufacturer/tradename
Millistak+®
parameter
1 bar max. differential pressure (15 psid) at 80 °C (Forward)
1 bar max. inlet pressure (15 psi) at 80 °C
2.1 bar max. differential pressure (30 psid) at 25 °C (Forward)
2.1 bar max. differential pressure (30 psid) at 25 °C (Reverse)
3.5 bar max. inlet pressure (50 psi) at 25 °C
H
32 cm (12.5 in.)
L
62 cm (24.2 in.)
filtration area
1.1 m2
impurities
<0.25 EU/mL bacterial endotoxins (LAL test, Aqueous extraction)
input
sample type liquid
conductivity
19.34-53.2 μS/cm at 25 °C (product water)
filter grade
X0HC
Looking for similar products? Visit Product Comparison Guide
General description
This X0HC media series delivers optimal performance through a gradient density matri, ideal for secondary clarification post depth filtration or centrifugation. Can also be utilized in downstream processing steps to protect chromatography columns. applications. Millistak+® Pod filters are available in three distinct series of media grades in order to meet your specific needs - DE, CE and HC.
The Millistak+® Pod system is ideal for a wide variety of primary and secondary clarification applications, including cell cultures, yeast and E. coli lysates post centrifuge, E. coli refolds, media, vaccines, plasma proteins and sera.
Easy to Use
With the compact, modular design of our Millistak+® new Pod system, you can increase productivity and shorten cycle times.
Installation and setup of the Pod system is simple and straightforward. The unique design of the disposable adapters and disposable manifolds makes it easy to connect the Pod filters to rest of the unit operations in the process. The self-contained and disposable nature of the system protects operators from exposure to biohazards and eliminates maintenance as well as cleaning validation requirements.
Application
Features and Benefits
- Low hold-up volume for greater product yield
- Broad range of media types offered in single and multilayer products
- Millistak+® HC dual-action media improves prefiltration and compresses clarification
- Flexible, modular format offers scalability from 5 to 12,000 liters or more
- Patented disposable design eliminates need for housing, CIP or cleaning validation
- Self-contained Pod filters protect operators from exposure to biohazards
- Robust construction is easy to use and set up
- Smaller footprint facilitates use in tight spaces
Specifications
Sodium: <14.1 mg/ft2
Calcium: <5.3 mg/ft2
Magnesium: <3.9 mg/ft2
Mercury: <0.001mg/ft2
Chromium: ≤0.001 mg/ft2
Aluminum: <0.084 mg/ft2
Potassium: <4.2mg/ft2
Lead: <0.001 mg/ft2
Arsenic: <0.009mg/ft2
Cobalt: ≤0.001 mg/ft2
Iron: <0.064 mg/ft2
Manganese: ≤ 0.124 mg/ft2
Titanium: <0.002 mg/ft2
Zinc: <0.046 mg/ft2
Nickel: <0.013
Copper: <0.009
Preparation Note
1 autoclave cycle of 60 min @ 123 ºC
Analysis Note
Legal Information
What is the Emprove® Program?
The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:
Free of charge - Sign in to download
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service