Skip to Content
Merck
All Photos(1)

Key Documents

MSSCNVX00

Millipore

Multiscreen® 96 well Plate, solid bottom

V-bottom clear polystyrene plate, non-sterile, pkg of 100 plates

Synonym(s):

MultiScreen® 96-well plate

Sign Into View Organizational & Contract Pricing

Select a Size

100 EACH
1.870,00 kr.

1.870,00 kr.


Estimated to ship on08 September 2025Details



Select a Size

Change View
100 EACH
1.870,00 kr.

About This Item

UNSPSC Code:
41122102
NACRES:
NB.15
material:
V-bottom clear polystyrene plate
size:
96 well
sterility:
non-sterile

1.870,00 kr.


Estimated to ship on08 September 2025Details


material

V-bottom clear polystyrene plate

Quality Level

sterility

non-sterile

product line

MultiScreen®

packaging

pkg of 100 plates

plate size

96 wells

size

96 well

Looking for similar products? Visit Product Comparison Guide

General description

MultiScreen® 96-well polystyrene microplates meet industry standard dimensions for use in common biochemical assay platforms, including automated liquid handling, microplate readers, and other microplate compatible devices. Polystyrene microplates are appropriate for a variety of biochemical assays in addition to sample preparation and storage.

Physical form

Color Code:Clear

Other Notes

Automation Compatible:Yes

Legal Information

MULTISCREEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service