Skip to Content
Merck
All Photos(1)

Key Documents

MHWP037A0

Millipore

Aerosol Monitor, 37 mm, mixed cellulose esters, 0.45 µm, white, with pad

Microscopic particle analysis and sample collection

Sign Into View Organizational & Contract Pricing

Select a Size

50 PKG
1.760,00 kr.

1.760,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
50 PKG
1.760,00 kr.

About This Item

UNSPSC Code:
41116116
eCl@ss:
32161002
NACRES:
NB.24

1.760,00 kr.


Please contact Customer Service for Availability

material

cellulose membrane
mixed cellulose esters (MCE) membrane
plain filter
polystyrene
white filter

Quality Level

sterility

non-sterile

filter diam.

37 mm

filtration area

9 cm2

pore size

0.45 μm pore size

fitting

female Luer inlet slip
female Luer outlet slip

application(s)

environmental

shipped in

ambient

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Sol. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service