Skip to Content
Merck
All Photos(1)

Key Documents

MABX7621

Sigma-Aldrich

Anti- Creatine Kinase-MB Isoenzyme (CKMB), clone 1

clone 1, from mouse

Synonym(s):

CKMB

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
7.470,00 kr.

7.470,00 kr.


Estimated to ship on14 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 MG
7.470,00 kr.

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

7.470,00 kr.


Estimated to ship on14 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody product type

primary antibodies

clone

1, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG1

Gene Information

human ... CKM(1158)

General description

Antibody recognises CKMB which is an isoform of Creatine Kinase present in heart muscle cells. CKMB is a well established cardiac biomarker, measurement of the CKMB levels in the blood are routinely used to assist in the diagnosis of a myocardial infarction.

Immunogen

CKMB

Application

Confirmed matched pairing. Capture Antibody MABX7621-1KC; Detector Antibody MABX7622-1KC
ELISA;Western Blotting
Research Category
Cancer

Physical form

50 mM NaCl 10 mM PBS pH 8.0 0.1 % sodium azide (NaN₃)
Format: Purified
Protein A purified

Storage and Stability

Stable for 12 months at 2-8ºC from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service