Skip to Content
Merck
All Photos(1)

Key Documents

MABT1528

Sigma-Aldrich

Anti-Claudin 5 Antibody, clone 2B12

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
969,00 kr.
100 μL
2.500,00 kr.

969,00 kr.


Estimated to ship on03 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 μL
969,00 kr.
100 μL
2.500,00 kr.

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

969,00 kr.


Estimated to ship on03 April 2025Details


Request a Bulk Order

antibody form

purified antibody

clone

2B12
monoclonal

purified by

using protein G

species reactivity

human, human, mouse

concentration

(Please refer to lot specific datasheet.)

technique(s)

ELISA: suitable
flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

UniProt accession no.

Mode of action

| inhibits

General description

An artificial membrane protein consisting of the extracellular region of human Claudin 5.

Specificity

Clone 2B12 is a mouse monoclonal antibody that detects human Claudin 5. It targets an epitope within nine amino acids in the second extracellular domain.

Application

Epitope: extracellular domain

Quality

Evaluated by Flow Cytometry in human umbilical vein endothelial cells (HUVEC).

Flow Cytometry Analysis: 1 µg of this antibody detected Claudin 5 one million human umbilical vein endothelial cells (HUVEC).

Physical properties

23.15 kDa calculated.

Physical form

Purified mouse monoclonal antibody IgG2a in PBS without azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service