Skip to Content
Merck
All Photos(1)

Documents

MABS2279

Sigma-Aldrich

Anti-Serpin B13 Antibody, clone B34

Synonym(s):

Serpin family B member 13

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

B34, monoclonal

mol wt

calculated mol wt 44.32 kDa

purified by

using protein G

species reactivity

mouse

packaging

antibody small pack of 100 μg

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

isotype

IgG2aκ

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

Related Categories

General description

Serpin B13 (UniProt: Q8CDC0; also known as Serpin family B member 13 ) is encoded by the Serpinb13 gene (Gene ID: 241196) in murine species. Serpin B13 is a cytoplasmic protein that plays a role in the proliferation or differentiation of keratinocytes. Two isoforms of Serpin B13 have been described that are generated by alternative splicing. Serpin B13 acts as a serine proteinase inhibitor and studies have shown a strong induction of serpin B13 in the pancreas of mice during the first few weeks of life. Cathepsin L and K appear to be the targets of serpin B13 action. Serpin B13 is expressed in pancreas with low levels of expression in pancreatic islets and larger expression in the exocrine pancreas. Secretion of anti-serpin B13 autoantibodies in young diabetes-prone non-obese diabetic (NOD) mice is shown to be associated with reduced inflammation in pancreatic islets and a slower progression to autoimmune diabetes. Anti-serpin B13 autoantibodies are shown to protect NOD mice against diabetes before 16 weeks of age. Reduced expression of serpin B13 autoantibodies is associated with development of type I diabetes in human before 5 years of age. Mice injected with monoclonal anti-serpin B13 display few inflammatory cells in the islets and rapid recovery from recent onset diabetes. (Ref.: Kryvalap, Y., et al. (2021). Sci. Transl. Med. 13(588); eabf1587; Czyzyk, J. et al. (2012). J. Immunol. 188(12); 6319-6327).

Specificity

Clone B34 is a mouse monoclonal antibody that detects mouse Serpin B13.

Immunogen

Recombinant full-length mouse Serpin B13.

Application

Quality Control Testing

Evaluated by Immunohistochemistry (Paraffin) in mouse skin tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution of this antibody detected Serpin B13 in mouse skin tissue sections.

Tested Applications

Immunohistochemistry Applications: A representative lot detected Serpin B13 in Immunohistochemistry applications (Kryvalap, Y., et al. (2021). Sci Transl Med.13(588):eabf1587).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Serpin B13, clone B34, Cat. No. MABS2279, is a mouse monoclonal antibody that detects Serpin B13 and is tested for use in Immunohistochemistry (Paraffin).

Physical form

Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service