Skip to Content
Merck
All Photos(1)

Documents

MABS1704

Sigma-Aldrich

Anti-Potassium Channel Kir6.2 Antibody, clone 10C1.1

clone 10C1.1, from mouse

Synonym(s):

ATP-sensitive inward rectifier potassium channel 11, IKATP, Inward rectifier K(+) channel Kir6.2, Potassium channel, inwardly rectifying subfamily J member 11

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

10C1.1, monoclonal

species reactivity

human, rat

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... KCNJ11(3767)

General description

ATP-sensitive inward rectifier potassium channel 11 (UniProt Q14654; also known as IKATP, Inward rectifier K(+) channel Kir6.2, Potassium channel, inwardly rectifying subfamily J member 11) is encoded by the KCNJ11 (also known as BIR, HHF2, MODY13, PNDM, TNDM3) gene (Gene ID 3767) in human. Inward rectifier K+ channels (Kirs) modulate cell membrane potential and are classified as K+ transport channels (Kir1.x, Kir4.x, Kir5.x, Kir7.x), classic Kir (Kir2.x), G-protein-gated Kirs (Kir2.x and Kir3.x), and ATP-sensitive Kirs (Kir6.x). Kirs mediate diverse physiological functions of both excitable and nonexcitable epithelial cells, such as cell adhesion-migration, volume regulation, apoptosis, and proliferation. ATP-sensitive potassium (K-ATP) channels are hetero-octamers composed of pore-forming Kir6.x (6.1 or 6.2) subunits and sulfonylurea receptor (SUR1 or SUR2) regulatory subunits. K-ATP channels are regulated by intracellular ATP and ADP concentrations and are highly expressed in cardiomyocytes, where they play crucial roles in ischemic preconditioning (IPC). Potassium channel openers mimic the cardioprotection, while K-ATP channel blocker or Kir6.2-knockout is shown to abolish the effect of IPC in mice. Human Kir6.2 is a 390-amino acid multi-pass membrane (a.a. 69-93, 117-128, 145-166) protein that contains two small extraceullar regions (a.a. 94-116 and 136-144) and a pore-forming domain (a.a. 117-128), having both its N- and C-terminal exposed intracellularly (a.a. 1-68 and a.a. 167-390).

Specificity

Clone 10C1.1 targets human Kir6.2 N-terminal cytoplasmic region not present in the alternatively spliced isoform 2.

Immunogen

Epitope: N-terminal cytoplasmic domain.
KLH-conjugated linear peptide corresponding an N-terminal cytoplasmic domain sequence of human potassium channel Kir6.2.

Application

Research Category
Signaling
Research Sub Category
Ion Channels & Transporters
This Anti-Potassium Channel Kir6.2 Antibody, clone 10C1.1 is validated for use in Western Blotting for the detection of Potassium Channel Kir6.2.
Western Blotting Analysis: A 1:1000 dilution from a representative lot detected potassium channel Kir6.2 in 10 µg of rat pancreas tissue lysate.

Quality

Evaluated by Western Blotting in human pancreas tissue lysate.

Western Blotting Analysis: A 1:1000 dilution of this antibody detected potassium channel Kir6.2 in 10 µg of human pancreas tissue lysate.

Target description

~45 kDa observed. 43.54 kDa (human) and 43.61 kDa (rat) calculated.

Physical form

Format: Purified
Protein G purified.
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service