Skip to Content
Merck
All Photos(1)

Key Documents

MABS1249

Sigma-Aldrich

Anti-SCAP Antibody, clone 9D5

clone 9D5, from mouse

Synonym(s):

Sterol regulatory element-binding protein cleavage-activating protein, SCAP, SREBP cleavage-activating protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
2.750,00 kr.

2.750,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
2.750,00 kr.

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

2.750,00 kr.


Please contact Customer Service for Availability

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9D5, monoclonal

species reactivity

hamster

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

hamster ... Scap(100689048)

General description

Sterol regulatory element-binding protein cleavage-activating protein (UniProt P97260; also known as SCAP, SREBP cleavage-activating protein) is encoded by the SCAP gene (Gene ID 100689048) in hamster species. SCAP mediates the activation of membrane-bound transcription factors SREBPs (sterol-regulatory element binding proteins) for transcribing genes encoding cholesterol biosynthetic enzymes. In sterol-deprived cells, SCAP escorts SREBPs from ER to Golgi, where active fragments of SREBPs are released from membrane by proteolysis. Sterol accumulation inuduces a negative regulatory mechanism, where SCAP becomes bound by Insigs and trapped in the ER, preventing further delivery of SREBPs to the Golgi for proteolytic release. SCAP spans the ER membrane eight times (a.a. 19-39, 280-300, 313-333, 345-365, 402-422, 424-444, 519-539, 709-729) with
4 lumenal and 5 cytoplasmic regions, having both its N- and C-terminal ends at the cytoplasmic side (a.a. 1-18 & 730-1276). The C-terminal domain of SCAP mediates association with SREBPs, while transmembrane helices 2–6 comprise the sterol-sensing domain that mediates sterol-induced binding of SCAP to Insigs. Mutations within the sterol-sensing region disrupt Insig binding and prevent sterol-mediated ER retention of SCAP-SREBP.

Specificity

Clone 9D5 detected the target band in lysate and membrane extract from CHO-K1 cells, but not from the SCAP-deficient SRD-13A CHO cell line.

Immunogen

Epitope: The fourth lumenal domain.
His-tagged recombinant fragment corresponding to the fourth lumenal domain of hamster SCAP.

Application

Anti-SCAP Antibody, clone 9D5 is an antibody against SCAP for use in Western Blotting, Immunocytochemistry.
Western Blotting Analysis: 5 µg/mL from a representative lot detected SCAP in 25 µg of whole cell lysate and 50 µg of membrane extract from CHO-K1 cells, but not from the SCAP-deficient SRD-13A CHO cells (Courtesy of Linda Donnelly, Department of Molecular Genetics, UT Southwestern Medical Center, Dallas, TX, USA).
Western Blotting Analysis: A representative lot detected exogenously expressed SCAP co-immunoprecipitated with the exogenously expressed Insig-1 and Insig-2 upon 25-hydroxycholesterol (25-HC) treatment of a SCAP-deficient CHO cell line SRD-13A that had been transfected to co-express SCAP with either myc-tagged Insig-1 or Insig-2 (Lee, P.C., and DeBose-Boyd, R.A. (2010). J. Lipid Res. 51(1):192-201).
Western Blotting Analysis: Representative lots detected similar level of SCAP in the membrane extracts from CHO cells with or without SREBP processing inhibitor 25-hydroxycholesterol (25-HC) treatment regardless whether the cells′ 25-HC-resistant status (Lee, P.C., and DeBose-Boyd, R.A. (2010). J. Lipid Res. 51(1):192-201; Lee, P.C., et al. (2007). J. Lipid Res. 48(9):1944-1954).
Western Blotting Analysis: A representative lot detected the endogenous SCAP immunoprecipitated from CHO cell lysate by a polyclonal SCAP antibody (Sakai, J., et al. (1997). J. Biol. Chem. 272(32):20213-20221).
Immunocytochemistry Analysis: A representative lot localized the overexpressed hamster SCAP at ER and nuclear envelope by fluorescent immunocytochemistry staining of 3% paraformaldehyde-fixed, 0.01% saponin-permeabilized CHO stable transfectants (Sakai, J., et al. (1997). J. Biol. Chem. 272(32):20213-20221).

Quality

Identity Confirmation by Isotyping Test.

Isotyping Analysis: The identity of this monoclonal antibody is confirmed by isotyping test to be IgG2bκ.

Target description

~150 kDa observed. Target band size appears larger than the calculated molecular weight of 139.5 kDa due to glycosylation. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service