Skip to Content
Merck
All Photos(3)

Key Documents

MABN691

Sigma-Aldrich

Anti-GPI Antibody, clone 1B7D7

ascites fluid, clone 1B7D7, from mouse

Synonym(s):

Glucose-6-phosphate isomerase, Autocrine motility factor, Neuroleukin, Phosphoglucose isomerase, Phosphohexose isomerase, Sperm antigen 36

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
3.090,00 kr.

3.090,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
3.090,00 kr.

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

3.090,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

1B7D7, monoclonal

species reactivity

rat, human

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NLK(51701)

General description

Glucose-6-phosphate isomerase (GPI) alternatively known as Autocrine motility factor (AMF), Neuroleukin (NLK), Phosphoglucose isomerase (PGI), Phosphohexose isomerase (PHI) or Sperm antigen 36 (SA-36) and encoded by the gene name GPI is major enzyme in glycolysis and gluconeogenesis. Glucose-6-phosphate isomerase rearranges glucose-6-phosphate (from the phosphorylation of glucose via hexokinases) into fructose-6-phosphate (F6P) as the second step in glycolysis. Interestingly however, Glucose-6-phosphate isomerase plays an entirely different sort of role outside of the cell than when it is inside being part of glycolysis. When secreted, Glucose-6-phosphate isomerase acts as an autocrine motility factor for various cancers and is critical for metastasis, while when secreted in nervous tissue it acts as a neurotrophic factor for sensory and spinal neurons and in lymphocytes it acts as a lymphokine upon T cell ligand binding and stimulates B cells to secrete immunoglobulin. Mutations in Glucose-6-phosphate isomerase are the second most frequent cause of inherited glycolytic-enzymopathy in humans. This autosomal recessive disorder is characterized by anon-spherocytic anemia of variable severity which can present with neuromuscular dysfunctions defined by muscle weakness and mental retardation. EMD-Millipore’s Anti-Glucose-6-phosphate isomerase monoclonal antibody has been tested in western blot on recombinant protein as well as cell lysates from HepG2, SMMC-7721 and rat liver tissue and in paraffin embedded immunohistochemistry on human kidney and fluorescent immunocytochemistry on L-O2 cells in culture.

Immunogen

Purified recombinant fragment of human GPI expressed in E. Coli.

Application

Anti-GPI Antibody, clone 1B7D7 is a highly specific mouse monoclonal antibody, that targets Glucose-6-phosphate isomerase & has been tested in western blotting, IHC & Immunofluorescence.
Immunohistochemistry Analysis: A 1:200-1,000 dilution from a representative lot detected GPI in human kidney tissue.

Immunofluorescent Analysis: A 1:200-1,000 dilution from a representative lot detected GPI in L-02 cells.

Optimal working dilutions must be determined by end user.

Quality

Evaluated by Western Blotting in HepG2, SMMC-7721, and rat liver lysates.

Western Blotting Analysis: A 1:500-2,000 dilution of this antibody detected GPI in HepG2, SMMC-7721, and rat liver lysates.

Target description

~56 kDa observed. Uncharacterized bands may appear in some lysate(s).

Analysis Note

Control
HepG2, SMMC-7721, and rat liver lysates

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service