Skip to Content
Merck
All Photos(2)

Key Documents

MABN2266

Sigma-Aldrich

Anti-ARID1B Antibody, clone 3D9.1

clone 3D9.1, from mouse

Synonym(s):

AT-rich interactive domain-containing protein 1B, ARID domain-containing protein 1B, BRG1-associated factor 250b, BAF250B, BRG1-binding protein hELD/OSA1, Osa homolog 2, hOsa2, p250R

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
2.830,00 kr.

2.830,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
2.830,00 kr.

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

2.830,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3D9.1, monoclonal

species reactivity

human, mouse

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... ARID1B(57492)

General description

AT-rich interactive domain-containing protein 1B (UniProt: Q8NFD5; also known as ARID domain-containing protein 1B, BRG1-associated factor 250b, BAF250B, BRG1-binding protein hELD/OSA1, Osa homolog 2, hOsa2, p250R) is encoded by the ARID1B (also known as BAF250B, DAN15, KIAA1235, OSA2) gene (Gene ID: 57492) in human. ARID domain-containing protein 1B (ARID1B) is widely expressed with high levels in heart, skeletal muscle, and kidney. Four isoform of ARID1B have been reported that are produced by alternative splicing. Its ARID domain is localized to amino acids 1053-1144. ARID1B is a component of the SWI/SNF chromatin remodeling complex and may play a role in cell-cycle activation. It is involved in transcriptional activation and repression of select genes by chromatin remodeling (alteration of DNA-nucleosome topology). ARID1B and ARID1A display about 60% homology, but have been reported to have opposing functions in cell cycle arrest and are mutually exclusive since individual SWI/SNF chromatin remodeling complexes can contain either ARID1A or ARID1B, but not both. ARID1B belongs to the neural progenitors-specific chromatin remodeling complex (npBAF complex) and the neuron-specific chromatin remodeling complex (nBAF complex). During neural development a switch from a stem/progenitor to a post-mitotic chromatin remodeling mechanism occurs as neurons exit the cell cycle and become committed to their adult state. Mutations in ARID1B gene are linked to the development of Coffin-Siris syndrome 1, a congenital multiple malformation syndrome with intellectual disability, coarse facial features, and malformations of the cardiac, gastrointestinal, genitourinary, and/or central nervous systems.

Specificity

Clone 3D9.1 detects AT-rich interactive domain-containing protein 1B (ARID1B) in human and murine cells. It targets an epitope within 138 amino acids from the N-terminal half.

Immunogen

GST/His-tagged recombinant fragment corresponding to 138 amino acids from the N-terminal half of human AT-rich interactive domain-containing protein 1B.

Application

Anti-ARID1B, clone 3D9.1, Cat. No. MABN2266, is a highly specific mouse monoclonal antibody that targets AT-rich interactive domain-containing protein 1B and has been tested for use in Immunocytochemistry and Western Blotting.
Immunocytochemistry Analysis: A 1:250 dilution from a representative lot detected ARID1B in HeLa, A431, HUVEC and NIH/3T3 cells.

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected ARID1B in 10 µg of HeLa cell lysate.

Target description

~300 kDa observed; 236.12 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service