Recommended Products
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
6B10.2, monoclonal
mol wt
calculated mol wt 18.70 kDa
observed mol wt ~18 kDa
species reactivity
mouse, human
packaging
antibody small pack of 100 μL
isotype
IgG1κ
UniProt accession no.
shipped in
dry ice
storage temp.
-10 to -25°C
target post-translational modification
unmodified
Gene Information
human ... CD247(919)
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in Jurkat cell lysate.
Western Blotting Analysis: A 1:500 dilution of this antibody detected CD3 zeta (CD247) in Jurkat cell lysate.
Tested Applications
Flow Cytometry Analysis: A representative lot detected CD3 zeta (CD247) in Flow Cytometry applications (Machado Diaz, A.C., et. al. (2012). Arthritis. 2012:943156).
Immunoprecipitation Analysis: A representative lot immunoprecipitated CD3 zeta (CD247) in Immunoprecipitation applications (van Oers, N.S., et. al. (1995). J Exp Med. 182(5):1585-90).
Immunohistochemistry Applications: A representative lot detected CD3 zeta (CD247) in Immunohistochemistry applications (Balkhi, M.Y., et. al. (2018). iScience. 2:105-122).
Immunofluorescence Analysis: A representative lot detected CD3 zeta (CD247) in Immunofluorescence applications (Lee, J., et. al. (2020). NPJ Biofilms Microbiomes. 6(1):49).
Western Blotting Analysis: A representative lot detected CD3 zeta (CD247) in Western Blotting applications (Aki, D., et. al. (2018). Nat Immunol. 19(7):766-775; van Oers, N.S., et. al. (1995). J Exp Med. 182(5):1585-90).
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service