Skip to Content
Merck
All Photos(1)

Key Documents

MABE914

Sigma-Aldrich

Anti-Mi-2β Antibody, clone 16G4

clone 16G4, from mouse

Synonym(s):

Chromodomain-helicase-DNA-binding protein 4, CHD-4, ATP-dependent helicase CHD4, Mi-2 autoantigen 218 kDa protein, Mi2-beta

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
2.420,00 kr.

2.420,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
2.420,00 kr.

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

2.420,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

16G4, monoclonal

species reactivity

human

technique(s)

ChIP: suitable (ChIP-seq)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CHD4(1108)

General description

CHD-4, often referred to as Mi2-beta, is a central component of the nucleosome remodeling and histone deacetylase (NuRD) repressor complex. CHD-4 has been shown to interact with TRIM27, HDAC1, Histone deacetylase 2, MTA2, SATB1 and Ataxia telangiectasia and Rad3 related. This protein also interacts with KLF1. Sumoylation of KLF1 leads to transcriptional repression of CHD-4.

Immunogen

Recombinant protein corresponding to Human Mi-2β.

Application

Chromatin Immunoprecipitation (ChIP): A representative lot of this antibody was used for ChIP. See Zhang et al. (2012).

ChIP-seq: representative lot of this antibody was used for ChIP-seq. See Zhang et al. (2012).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology
This Anti-Mi-2β Antibody, clone 16G4 is validated for use in Western Blotting and Chromatin Immunoprecipitation (ChIP) and ChIP-seq for the detection of Mi-2β.

Quality

Evaluated by Western Blotting in a Jurkat cell lysate.

Western Blotting Analysis: A 1 µg/mL dilution of this antibody detected Mi-2β in a Jurkat cell lysate.

Target description

~220 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ supernatant in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service