Skip to Content
Merck
All Photos(2)

Key Documents

MABE222

Sigma-Aldrich

Anti-JMJD2A Antibody, clone 8B7.1, rabbit monoclonal

clone 8B7.1, from rabbit

Synonym(s):

JMJD, Lysine-specific demethylase 4A, JmjC domain-containing histone demethylation protein 3A, Jumonji domain-containing protein 2A

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
3.030,00 kr.

3.030,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
3.030,00 kr.

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

3.030,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

8B7.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KDM4A(9682)

General description

Jumonji domain-containing protein 2A (JMJD2A), or KDM4A, is a ubiquitously expressed demethylase enzyme, which belongs to the JMJD2 family. Like other members of this family, JMJD2A contains multiple domains: two Tudor domains, two PHD-type zinc fingers, one JmjC domain, and one JmjN domain. Little is known about the function of most of these domains; however, they are believed to be involved in transcriptional regulation. JMJD2A functions as a histone demethylase with specific activity against trimethylated Histone H3 Lysine residues Lys-9 and Lys-36. It has no effect on any other lysine residues, or on mono- and dimethylated Lys-9 and Lys-36. The demethylation of these residues induces the production of succinate and formaldehyde. Studies have shown that JMJD2A recruits NCOR1 and histone deacetylases, causing the respective transcriptional repression of E2Fand ASCL2 responsive promoters.

Immunogen

KLH-conjugated linear peptide corresponding to human JMJD2A.

Application

Immunohistochemistry Analysis: A 1:100 dilution from a representative lot detected JMJD2A in human colorectal adenocarcinoma cells.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histone Modifying Proteins
This Anti-JMJD2A Antibody, clone 8B7.1 is validated for use in Western Blotting, IHC for the detection of JMJD2A.

Quality

Evaluated by Western Blot in U2OS nuclear extract.

Western Blot Analysis: A 1:1,000 dilution of this antibody detected JMJD2A in 10 µg of U2OS nuclear extract.

Target description

~120 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
U2OS nuclear extract

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service