Skip to Content
Merck
All Photos(4)

Key Documents

MABE1114

Sigma-Aldrich

Anti-CENP-C Antibody, clone AL61A

clone AL61A, from mouse

Synonym(s):

Centromere protein C, CENP-C, CENP-C 1, Centromere autoantigen C, Centromere protein C 1, Interphase centromere complex protein 7

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
2.730,00 kr.

2.730,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
2.730,00 kr.

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

2.730,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

AL61A, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CENPC(1060)

General description

Centromere protein C (UniProt Q03188; also known as CENP-C, CENP-C 1, Centromere autoantigen C, Centromere protein C 1, Interphase centromere complex protein 7) is encoded by the CENPC (also known as CENPC1, ICEN7) gene (Gene ID 1060) in human. Kinetochores assemble on centromeres to bind spindle microtubules and mediate chromosome segregation during cell division. CENP-C is a subunit of constitutive centromere-associated network (CCAN) that acts as the centromere-kinetochore interface. CCAN is a large complex composed of at least 16 different centromeric proteins (CENPs). CENP-C plays an important role in kinetochore assembly downstream of CENP-A. The PEST domain in the N-terminal half of CENP-C interacts directly with the CCAN subcomplex CENP-HIKM (CENP-H, CENP-K, CENP-I, and CENP-M) and CENP-C depletion is shown to casue mislocalization of CENP-T and the CENP-HIKM complex subunits in HeLa cells.

Specificity

Expected to react with both spliced isoforms of human CENP-C reported by UniProt (Q03188).

Immunogen

Epitope: N-terminal.
GST-tagged recombinant human CENP-C N-terminal fragment.

Application

Anti-CENP-C Antibody, clone AL61A is an antibody against CENP-C for use in Immunocytochemistry, Immunoprecipitation.
Immunoprecipitation Analysis: Prior to antibody purification, clone AL61A hybridoma culture supernatant immunoprecipitated CENP-C from HeLa nuclear extract (Courtesy of Dr. Giovanna Roncador, Spanish National Cancer Research Centre (CNIO), Madrid, Spain).
Immunocytochemistry Analysis: Prior to antibody purification, clone AL61A hybridoma culture supernatant detected nuclear CENP-C puncta by fluorescent immunocytochemistry staining of permeabilized HeLa and Jurkat cells (Courtesy of Dr. Giovanna Roncador, Spanish National Cancer Research Centre (CNIO), Madrid, Spain).
Immunocytochemistry Analysis: Prior to antibody purification, clone AL61A hybridoma culture supernatant immunostained CENP-C-transfected, but not ZBTB32-transfected, HEK293T cells (Courtesy of Dr. Giovanna Roncador, Spanish National Cancer Research Centre (CNIO), Madrid, Spain).

Quality

Evaluated by Immunocytochemistry in Jurkat cells.

Immunocytochemistry Analysis: A 1:250 dilution of this antibody detected CENP-C in Jurkat cells.

Target description

106.8 kDa (isoform 1) and 61.54 kDa (isoform 2) calculated.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service