Skip to Content
Merck
All Photos(1)

Key Documents

MAB985

Sigma-Aldrich

Anti-Echovirus 4 Antibody, clone 431-4H-6H-3B

ascites fluid, clone 431-4H-6H-3B, Chemicon®

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
4.530,00 kr.

4.530,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
4.530,00 kr.

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.42

4.530,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

431-4H-6H-3B, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

isotype

IgG2a

shipped in

dry ice

Specificity

Reacts with Echovirus 4, both Pesasceck and Dutoit prototype strains. May also react with several strains of Echovirus 30.

Application

This Anti-Echovirus 4 Antibody, clone 431-4H-6H-3B is validated for use in IF for the detection of Echovirus 4.
When used in conjunction with Echovirus 30 MAB, MAB 985 can be used to identify Echovirus 4. Isolates positive with MAB 985 should be tested with Echovirus 30 MAB (MAB 974 or Cat. No. 3315) to verify its proper identification. If the isolate is positive with Echovirus 30 MAB or both Echovirus 30 and Echovirus 4 MABs then the isolate can be presumptively identified as Echovirus 30. If the isolate is only positive with Echovirus 4 MAB then the isolate can be presumptively identified as Echovirus 4.

Suggested starting dilution for indirect immunofluorescence is 1:1000. Dilute with buffer pH 7.8 containing protein or other stabilizing medium. Final working dilutions must be determined by end user.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service