Skip to Content
Merck
All Photos(1)

Key Documents

MAB8126

Sigma-Aldrich

Anti-Cytomegalovirus Antibody, late Antigen, clone 2D4.2

clone 2D4.2, Chemicon®, from mouse

Synonym(s):

CMV

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
3.450,00 kr.

3.450,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
3.450,00 kr.

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

3.450,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2D4.2, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2a

shipped in

wet ice

Specificity

Reacts with a late protein M.W. 47-55 kD. Can detect CMV infection 24 hours post-infection exhibiting a cytoplasmic staining which reduces peak intensity at >72 hours.

With CMV the antigens expressed at different times are listed as:



Immediate Early (alpha gene expression): those antigens expressed at 3-12 hrs post-infection generally involved in Transcription such as 72kD major phosphoprotein and a few other other antigens at 60 - 80kD.

Early Antigen (beta genes) a.k.a. Delayed Early or Intermediate Early: expressed at 12-24hrs post-infection. Generally enzymes and one virion structural gene preceding viral DNA synthesis.

Late Antigen (gamma genes): expressed at 36-48hrs post-infection. Generally structural proteins. Major protein = 55kD

Immunogen

Epitope: late antigen
Sucrose gradient purified CMV AD169 (ATCC).

Application

Anti-Cytomegalovirus Antibody, late antigen, clone 2D4.2 detects level of Cytomegalovirus & has been published & validated for use in IF, WB, IH(P).
IFA at 1:400-1:800 on acetone fixed cells.

Works on paraffin embedded tissue sections.

Dilute with buffer pH 7.4-7.6 to desired working volume.

For extensive dilution, protein containing or other stabilizing medium should be used.

Final working dilutions must be determined by end user.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral

Physical form

Format: Purified
Purified immunoglobulin. Liquid in 0.02M PB with 0.25M NaCl, pH=7.6, 0.1% Sodium Azide.

Storage and Stability

Maintain at +2-8°C for up to 12 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of models and detection methods for different forms of cytomegalovirus for the evaluation of viral inactivation agents.
Vasudevacharya Jayarama, Jennifer Marcello, Asa Ohagen, Veronica Gibaja et al.
Transfusion null
William E Lawson et al.
American journal of physiology. Lung cellular and molecular physiology, 294(6), L1119-L1126 (2008-04-09)
Recent evidence suggests that dysfunctional type II alveolar epithelial cells (AECs) contribute to the pathogenesis of idiopathic pulmonary fibrosis (IPF). Based on the hypothesis that disease-causing mutations in surfactant protein C (SFTPC) provide an important paradigm for studying IPF, we
Sylwia Libard et al.
PloS one, 9(9), e108861-e108861 (2014-10-01)
Human cytomegalovirus (HCMV) has been indicated being a significant oncomodulator. Recent reports have suggested that an antiviral treatment alters the outcome of a glioblastoma. We analysed the performance of commercial HCMV-antibodies applying the immunohistochemical (IHC) methods on brain sample obtained

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service