Skip to Content
Merck
All Photos(1)

Key Documents

MAB4619

Sigma-Aldrich

Anti-Bcl-XL Antibody, NT, clone 7D9

clone 7D9, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
3.780,00 kr.

3.780,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 μG
3.780,00 kr.

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

3.780,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

7D9, monoclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunofluorescence: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BCL2L1(598)

Related Categories

General description

Bcl-X consists of two isoforms, Bcl-XL (long), a 241 aa protein, and Bcl-XS (short), a 178 aa protein lacking a 63-aa domain that is well conserved among members of the Bcl-2 family. Bcl-XL represses cell death, while Bcl-XS favors cell death.Bcl-X has two isoforms, Bcl-XL (long) a 241 amino acid protein and Bcl-XS (short) a 178 amino acid protein lacking a 63 amino acid domain that is well conserved among members of the Bcl-2 family. Bcl-XL represses cell death, while Bcl-XS favors cell death.

Specificity

MAB4619 recognizes a protein of 27 kDa, identified as the Bcl-XL protein. Highly specific to Bcl-XL and shows no cross-reaction with Bcl-XS, Bcl-2alpha or Bax protein.

Immunogen

Synthetic peptide corresponding to amino acids 35-50 (TEAPEETEAERETPSA) of mouse Bcl-XL protein. This sequence is conserved at 15 of 16 residues in rat Bcl-XL.

Application

Immunofluorescence

Flow Cytometry

Optimal working dilutions must be determined by end user.
This Anti-Bcl-XL Antibody, N-terminus, clone 7D9 is validated for use in FC, IF for the detection of Bcl-XL.

Physical form

Format: Purified
Immunoglobulin purified by protein A affinity chromatography. Liquid in 10mM PBS, pH 7.4, with 0.2% BSA and 0.09% sodium azide.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yiqing Li et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(2), E209-E218 (2017-01-05)
Retinal ganglion cells (RGCs), the projection neurons of the eye, cannot regenerate their axons once the optic nerve has been injured and soon begin to die. Whereas RGC death and regenerative failure are widely viewed as being cell-autonomous or influenced

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service