Skip to Content
Merck
All Photos(1)

Documents

MAB4073

Sigma-Aldrich

Anti-CD44v6 Antibody, exon v6, clone VFF-18

clone VFF-18, Chemicon®, from mouse

Synonym(s):

Anti-CDW44, Anti-CSPG8, Anti-ECMR-III, Anti-HCELL, Anti-HUTCH-I, Anti-IN, Anti-LHR, Anti-MC56, Anti-MDU2, Anti-MDU3, Anti-MIC4, Anti-Pgp1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

VFF-18, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... CD44(960)

Related Categories

Specificity

Specific for an epitope encoded by exon v6 on the variant portion of human CD44.

Immunogen

Epitope: exon v6

Application

Immunohistochemistry on acetone fixed cryostat sections using the alkaline phosphatase-anti-alkaline-phosphatase (APAAP) or peroxidase anti-peroxidase (PAP) procedures. Antibody may be used at a concentration of 1 - 2 μg/mL.

Immunohistochemistry on paraffin embedded sections after microwave treatment (1 x 5 min). Antibody may be used at a concentration of 1 - 2 μg/mL.

Immunocytochemistry on cell smears.

Western blot

Flow cytometry: 1:20 dilution

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)
This Anti-CD44v6 Antibody, exon v6, clone VFF-18 is validated for use in FC, WB, IC, IH(P) for the detection of CD44v6.

Physical form

Format: Purified
Purified immunoglobulin. Liquid in PBS with no preservative.

Storage and Stability

Maintain frozen at -20°C in undiluted aliquots for up to 12 months from date of receipt. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Osteopontin enhances the expression and activity of MMP-2 via the SDF-1/CXCR4 axis in hepatocellular carcinoma cell lines.
Zhang, R; Pan, X; Huang, Z; Weber, GF; Zhang, G
Testing null
Karthiga Santhana Kumar et al.
SpringerPlus, 4, 19-19 (2015-01-28)
Medulloblastoma (MB) comprises four molecularly and genetically distinct subgroups of embryonal brain tumors that develop in the cerebellum. MB mostly affects infants and children and is difficult to treat because of frequent dissemination of tumor cells within the leptomeningeal space.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service