Skip to Content
Merck
All Photos(2)

Key Documents

MAB2605

Sigma-Aldrich

Anti-ATG5 Antibody, clone 177.19

clone 177.19, from mouse

Synonym(s):

APG5 autophagy 5-like, APG5 autophagy 5-like (S. cerevisiae), APG5 (autophagy 5, S. cerevisiae)-like, ATG5 autophagy related 5 homolog (S. cerevisiae), ATG5 autophagy related 5-like, Apoptosis-specific protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
3.210,00 kr.

3.210,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
3.210,00 kr.

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

3.210,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

177.19, monoclonal

species reactivity

human

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ATG5(9474)

General description

ATG5 is required for autophagy. It conjugates to ATG12 and associates with isolation membrane to form autophagosomes. The conjugate detaches from the membrane immediately before or after autophagosome formation is completed. Conjugation to ATG12 is essential for autophagy, but is not required for association with isolation membrane. ATG5 also plays an important role in the apoptotic process. Its expression is a relatively late event in the apoptotic process, occurring downstream of caspase activity

Specificity

Other species have not been tested.
The antibody reacts with the 50 kDa ATG5 protein.

Immunogen

Linear Peptide, aa: 8-318

Application

Immunoprecipitation: HeLa cell lysate
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-ATG5 Antibody, clone 177.19 is validated for use in WB, IP for the detection of ATG5.

Quality

Western Blot Analysis: 0.5 ug/mL of this antibody detected ATG5 on HeLa cell lysate.

Target description

50 kDa

Linkage

Replaces: MABC137

Physical form

Format: Purified
Protein G Purified
Purified in 0.1M Tris-Glycine (pH7.4) 150mM NaCl with 0.05% NaN3.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Western Blot and Immunoprecipitation: HeLa cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wen-Tsung Huang et al.
Frontiers in pharmacology, 8, 902-902 (2018-01-13)
Intrinsic or acquired resistance to hormone therapy is frequently reported in estrogen receptor positive (ER+) breast cancer patients. Even though dysregulations of histone deacetylases (HDACs) are known to promote cancer cells survival, the role of different HDACs in the induction
Isabel Pombo Grégoire et al.
PLoS pathogens, 7(12), e1002422-e1002422 (2011-12-17)
Autophagy is a conserved degradative pathway used as a host defense mechanism against intracellular pathogens. However, several viruses can evade or subvert autophagy to insure their own replication. Nevertheless, the molecular details of viral interaction with autophagy remain largely unknown.
Tzu-Yu Lin et al.
Autophagy, 16(7), 1296-1313 (2019-10-16)
BIRC5/Survivin is known as a dual cellular functions protein that directly regulates both apoptosis and mitosis in embryonic cells during embryogenesis and in cancer cells during tumorigenesis and tumor metastasis. However, BIRC5 has seldom been demonstrated as a direct macroautophagy/autophagy

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service