Skip to Content
Merck
All Photos(1)

Key Documents

MAB10223

Sigma-Aldrich

Anti-L. pneumophila Serogroup 1 Antibody, clone Lp1 1

ascites fluid, clone Lp1 MAB 1, Chemicon®

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
2.980,00 kr.

2.980,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
2.980,00 kr.

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.42

2.980,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

Lp1 MAB 1, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

isotype

IgG3

shipped in

dry ice

General description

Legionella pneumophila are Gram-negative bacteria which are associated with Legionnaires′ disease and Pontiac fever. At least 14 different serovars of L. pneumophila have been described as well as several other species being subdivided into a number of serovars.

Specificity

The antibody reacts with a surface antigen on L. pneumophila

Immunogen

Bacterial antigenic subgroup OLDA

Application

Anti-L. pneumophila Serogroup 1 Antibody, clone Lp1 1 is an antibody against L. pneumophila Serogroup 1 for use in IF.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral

Physical form

Ascites
Liquid ascites fluid containing no preservatives.

Storage and Stability

1 year at -20°C from date of shipment

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J R Joly et al.
Journal of clinical microbiology, 23(4), 768-771 (1986-04-01)
A panel of monoclonal antibodies to Legionella pneumophila serogroup 1 and a subclassification scheme were developed in a collaborative project among three laboratories. The seven most useful monoclonal antibodies were selected from three previously developed panels on the basis of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service