Skip to Content
Merck
All Photos(1)

Key Documents

MAB10213

Sigma-Aldrich

Anti-G1 Envelope Glycoprotein Antibody, clone 807.31

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.42

antibody form

ascites fluid

Quality Level

clone

807.31
monoclonal

purified by

(Ascites)

species reactivity

human, mouse

technique(s)

immunofluorescence: suitable

isotype

IgG1

General description

G1 envelope glycoprotein of La Crosse Virus

Specificity

G1 envelope glycoprotein of La Crosse Virus

Physical form

Liquid ascites fluid containing no preservatives.

Storage and Stability

1 year at -20°C from date of shipment

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service