Skip to Content
Merck
All Photos(1)

Key Documents

LUXGCS3EXWA

Millipore

MPAGE®® Lux Warranty

3-year extended warranty for LUXGCS (mPAGE® Lux Curing Station), purchase at time-of-sale

Synonym(s):

Lux, Warranty

Sign Into View Organizational & Contract Pricing

Select a Size

LUXGCS3EXWA
1.030,00 kr.

1.030,00 kr.


Available to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
LUXGCS3EXWA
1.030,00 kr.

About This Item

UNSPSC Code:
41105312
NACRES:
NA.32

1.030,00 kr.


Available to ship on31 March 2025Details


Request a Bulk Order

Product Name

3-year extended warranty A for mPAGE® Lux Curing Station, 3-year extended warranty for LUXGCS (mPAGE® Lux Curing Station), purchase at time-of-sale

product line

mPAGE®

technique(s)

SDS-PAGE: suitable
electrophoresis: suitable

compatibility

mPAGE® LUXGCS

General description

3-year extended warranty for LUXGCS (mPAGE® Lux Curing Station), purchase at time-of-sale. The LUXGCS comes with a one-year warranty. This product extends the warranty to cover a total of four years. The instrument will be replaced if broken during the warranty period.

Other Notes

This is not a stand-alone product. It can only be purchased with mPAGE® Lux Curing Station

Legal Information

MPAGE is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service