Skip to Content
Merck
All Photos(1)

Key Documents

KS-1L-BK

Millipore

Anti-M (clone LM110/140), FFMU

Anti-M from the clone LM110/140, Murine IgG, Intermediate for further manufacturing use

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116102
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

unpurified

clone

LM-110/140, monoclonal

form

liquid

packaging

pack of 1 L

concentration

0.5-10 mg/mL protein (Total Protein Range)

technique(s)

agglutination assay: suitable

pH

6.7-7.7

isotype

IgG

shipped in

ambient

storage temp.

2-8°C

General description

Standardized, concentrated cell culture supernatant, non-diafiltered, minimal pH buffering; For the formulation of Anti-M blood typing reagents or devices.Direct agglutinating antibody.The optimal pH of an Anti-M finished reagent made from this antibody is 8.4. At pH values below 8.0 the antibody will react with NN red cells.Contains 0.1% (w/v) Sodium Azide; Shelf Life: 24 months from date of filtration.

Application

Antibody intermediate for further manufacturing use.
Production of an in vitro blood typing product or device.

Legal Information

Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Partially processed reagent for further manufacture of non-injectable products only. The absence of all viral antigens has not been determined. No US Standard of Potency. Do not use if turbid. Contains 0.1%<nbsp/>(w/v) sodium azide.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service