Skip to Content
Merck
All Photos(2)

Documents

IMGDV002

Millipore

Immobilon® GO for Simple Immunodetection

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116010
eCl@ss:
42029053
NACRES:
NA.47

manufacturer/tradename

Immobilon®

technique(s)

dot blot: suitable
western blot: suitable

compatibility

for use with Immobilon Western HRP Substrates
for use with Nitrocellulose
for use with PVDF (Immobilon membranes)
for use with TBS-T with NFDM
for use with commercially available detection reagents

detection method

chemiluminescent
colorimetric
fluorometric

shipped in

ambient

storage temp.

room temp

General description

Immobilon® GO applies the principles of lateral flow assays for walk away processing of Western blots. Simply place your miniblot in the device and then add your wash and antibody solutions to the ports in the top. Return later to apply detection substrate and image your Western blot.

Application

For use on immunoblots, Western blots and dot blots.

Legal Information

Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service