HTS108M
ChemiSCREEN Membrane Preparation Recombinant Human 5-HT1B Serotonin Receptor
Human 5-HT1B GPCR membrane preparation for Radioligand binding Assays & GTPγS binding.
Synonym(s):
5-HT1B Receptor Membrane Prep, ChemiSCREEN Serotonin Kit, Human 5-HT1B Membrane Prep
Select a Size
5.200,00 kr.
Select a Size
About This Item
5.200,00 kr.
Recommended Products
biological source
human
Quality Level
recombinant
expressed in Chem-1 cells
manufacturer/tradename
ChemiScreen
Chemicon®
technique(s)
ligand binding assay: suitable (GTPγS)
radioligand binding assay (RLBA): suitable
NCBI accession no.
UniProt accession no.
shipped in
dry ice
General description
Application
Biochem/physiol Actions
Quality
10 µg/well | 20 µg/well | |
---|---|---|
Signal:Background | 12.4 | 10.5 |
Specific Binding (cpm) | 3210 | 3352 |
SPECIFICATIONS: 1 unit =10 μg
Bmax for [3H]-GR125743 binding: 3.7 pmol/mg protein
Kd for [3H]-GR125743 binding: ~10.5 nM
Specifications
RECOMMENDED ASSAY CONDITIONS: Membranes are mixed with radioactive ligand and unlabeled competitor (see Figures 1 and 2 for concentrations tested) in binding buffer in a nonbinding 96-well plate, and incubated for 1-2 h. Prior to filtration, an FC 96-well harvest plate (Millipore cat. # MAHF C1H) is coated with 0.33% polyethyleneimine for 30 min, then washed with 50mM Tris, pH 7.4. Binding reaction is transferred to the filter plate, and washed 3 times (1 mL per well per wash) with Wash Buffer. The plate is dried and counted.
Binding buffer: 50 mM Tris, pH 7.4, 10 mM MgCl2, 1 mM EDTA, filtered and stored at 4°C
Radioligand: [3H]-GR125743. (Amersham #:TRK-1046 )
Wash Buffer: 50 mM Tris, pH 7.4, filtered and stored at 4°C.
One package contains enough membranes for at least 200 assays (units), where a unit is the amount of membrane that will yield greater than 8-fold signal:background with 3H labeled GR125743 at 7 nM
Physical form
Packaging method: Membranes protein were adjusted to 1 mg/ml in 1 ml packaging buffer, rapidly frozen, and stored at -80°C.
Storage and Stability
Legal Information
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service