Skip to Content
Merck
All Photos(1)

Documents

HTS099M

Sigma-Aldrich

ChemiSCREEN Membrane Preparation Recombinant Human EP1 Prostanoid Receptor

Human EP1 GPCR membrane preparation for Radioligand binding Assays & GTPγS binding.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32161000
NACRES:
NA.41

biological source

human

Quality Level

recombinant

expressed in Chem-1 cells

manufacturer/tradename

ChemiScreen
Chemicon®

technique(s)

ligand binding assay: suitable (GTPγS)
radioligand binding assay (RLBA): suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

General description

Full-length human PTGER1 cDNA encoding EP1
Prostanoids are a series of arachidonic acid metabolites produced by the action of cyclooxygenase and subsequently by isomerases and synthases. Cells rapidly secrete prostanoids after synthesis, whereupon the prostanoids bind to a family of 8 GPCRs to exert their biological effects (Narumiya and FitzGerald, 2001). The prostaglandin PGE2 causes pain, vasodilation, immunosuppression of T cells, bone resorption and promotion of carcinogenesis. Four related GPCRs, EP1, EP2, EP3 and EP4, each bind to PGE2, but the different G protein coupling status of each receptor leads to distinct biological effects; EP1 couples primarily to Gq to mobilize intracellular calcium. EP1 appears to mediate the effects of PGE2 in promoting formation of precancerous lesions in animal models of colon cancer (Watanabe et al., 1999). In addition, EP1 has an inhibitory effect on stress-induced aggressive and risk-taking behaviors in mice (Matsuoka et al., 2005). Millipore′s EP1 membrane preparations are crude membrane preparations made from our proprietary stable recombinant cell lines to ensure high-level of GPCR surface expression; thus, they are ideal HTS tools for screening of agonists and antagonists of EP1. The membrane preparations exhibit a Kd of 14.5 nM for [3H]-PGE2. With 40 nM [3H]-PGE2, 10 μg/well EP1 Membrane Prep typically yields greater than 3-fold signal-to-background ratio.

Application

Human EP1 GPCR membrane preparation for Radioligand binding Assays & GTPγS binding.
Radioligand binding assay and GTPγS binding

Biochem/physiol Actions

GPCR Class: A
Protein Target: EP1
Target Sub-Family: Prostanoid

Quality

Table 1. Signal:background and specific binding values obtained in a competition binding assay with EP1 membrane prep.
10 µg/well
Signal:Background 4.3
Specific Binding (cpm) 724

SPECIFICATIONS: 1 unit = 10 μg membrane preparation
Bmax: 4.6 pmol/mg
Kd: 14.5 nM

Specifications

Inucbation Conditions
Membranes are mixed with radioactive ligand and unlabeled competitor (see Figures 1 and 2 for concentrations tested) in binding buffer in a nonbinding 96-well plate, and incubated for 1-2 h. Prior to filtration, an FC 96-well harvest plate (Millipore cat. # MAHF C1H) is coated with 0.33% polyethyleneimine for 30 min, then washed with 10mM MES, pH 6, 1mM EDTA, 10mM MnCl2. Binding reaction is transferred to the filter plate, and washed 3 times (1 mL per well per wash) with Wash Buffer. The plate is dried and counted.

Binding buffer: 10mM MES, pH 6, 1mM EDTA, 10mM MnCl2, filtered and stored at 4°C

Radioligand: [3H]- PGE2. (Perkin Elmer # NET-428 )

Wash Buffer: 10mM MES, pH 6, 1mM EDTA, 10mM MnCl2, filtered and stored at 4°C.

One package contains enough membranes for at least 200 assays (units), where a unit is the amount of membrane that will yield greater than 3-fold signal: background with 3H labeled PGE2.

Physical form

Liquid in packaging buffer: 50 mM Tris pH 7.4, 10% glycerol and 1% BSA with no preservatives.
Packaging method: Membrane protein was adjusted to the indicated concentration in packaging buffer, rapidly frozen, and stored at -80°C.

Storage and Stability

Store at –70°C. Product is stable for at least 6 months from the date of receipt when stored as directed. Do not freeze and thaw.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service