Skip to Content
Merck
All Photos(1)

Key Documents

HCYTB-60K-PX38

Millipore

MILLIPLEX® Human Cytokine/Chemokine/Growth Factor Panel B 38-Plex Premix

Synonym(s):

MILLIPLEX Human Cytokine/Chemokine/Growth Factor Panel B 38 Plex Premixed Magnetic Bead Panel, The Human Cytokine/Chemokine/Growth Factor Panel B bead-based multiplex panel

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
52.510,00 kr.

52.510,00 kr.


Product Availability: You will receive an order acknowledgement once the order has been entered in our system.


Select a Size

Change View
1 KIT
52.510,00 kr.

About This Item

UNSPSC Code:
12161503
NACRES:
NA.77

52.510,00 kr.


Product Availability: You will receive an order acknowledgement once the order has been entered in our system.

usage

 kit sufficient for 96 wells

Quality Level

species reactivity

human

manufacturer/tradename

Milliplex®

parameter

25 μL sample volume (per well)

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex® xMAP®)

shipped in

wet ice

storage temp.

2-8°C

General description

®®“Cytokine” is a general term used for a diverse group of small proteins, peptides or glycoproteins secreted by lymphocytes, monocytes, macrophages and other cells that regulate immune responses, hematopoiesis and lymphocyte development. Cytokines include interleukins, interferons, chemokines and other signaling molecules. Growth factors are extracellular polypeptides that have a positive effect on cell growth and proliferation. They can stimulate growth in a variety of different cell types. Growth factors and cytokines are similar in structure and the way of action. Individual cytokines or growth factors are produced by multiple cell types. This is different from hormones, which tend to be made by specialized glands. Each cytokine or growth factor acts through its own receptor on target cells to generate signaling pathways, and as a consequence to regulate biological processes. Some intracellular signaling components are shared between cytokines and growth factors. Expression of cytokines or growth factors and their receptors is highly regulated. De-regulation may contribute to many diseases such as infectious disease, autoimmune and chronic inflammatory disease, cardiovascular disease, metabolic syndrome, neurological disorders, and cancer. Cytokine and growth factor research plays a significant role in achieving a deeper understanding of the immune system and combating related diseases.The MILLIPLEX® portfolio offers the broadest selection of analytes across a wide range of disease states and species. Once the analytes of interest have been identified, you can rely on the quality that we build into each kit to produce results you can trust. In addition to the assay characteristics listed in the protocol, other performance criteria evaluated during the verification process include: cross-reactivity, dilution linearity, kit stability, and sample behavior (for example, detectability and stability).In addition, each panel and kit meet stringent manufacturing criteria to ensure batch to-batch reproducibility. The MILLIPLEX® Human Cytokine/Chemokine/Growth Factor Panel B Magnetic Bead Panel enables you to focus on the therapeutic potential of cytokines and the modulation of cytokine expression. Coupled with the Luminex® xMAP® platform in a magnetic bead format, you receive the advantage of ideal speed and sensitivity, allowing quantitative multiplex detection of dozens of analytes simultaneously, which can dramatically improve productivity.

Research category: Inflammation & Immunology

Research sub-category:

  • Inflammation & Autoimmune Mechanisms
  • Inflammation

Specificity

There was no or negligible cross-reactivity between the antibodies for an analyte and any of the other analytes in this panel.

Application

The MILLIPLEX® Human Cytokine/Chemokine/Growth Factor Panel B Magnetic Bead 38-Plex Premixed Panel is a fixed 38-plex kit to be used for the simultaneous quantification of any or all of the following analytes in serum or plasma samples and tissue/cell lysate and culture supernatant samples: 6Ckine, sCD137, sFAS, sFASL, APRIL, BAFF, BCA-1, CTACK, ENA-78, Eotaxin-2, Eotaxin-3, Granzyme A, Granzyme B, HMGB1, I-309, IFNβ, IFNω, IL-16, IL-20, IL-21, IL-23, IL-28A, IL-29, IL-31, IL-33, I-TAC, LIF, MCP-2, MCP-4, MIP-1δ, MIP-3α, Perforin, SCF, SDF-1, TARC, TPO, TRAIL, TSLP.

Features and Benefits

Design your multiplex kit by choosing available analytes within this panel.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Cytokine multiplex assays allow researchers to easily investigate the immune system. The MILLIPLEX® Human Cytokine/Chemokine/Growth Factor Multiplex Panels A and B are multiplex assays for cytokine detection that each offer a unique combination of 48 analytes that can be simultaneously analyzed in a small sample volume.

MILLIPLEX® Human Cytokine/Chemokine/Growth Factor Panels A and B offer customizable options for analyzing nearly 100 cytokines.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service