Skip to Content
Merck
All Photos(1)

Key Documents

HAEP047AW

Millipore

6mm Hydrophobic Edge Membrane 0.45µm 47mm white plain autoclave packed

Filling machine operations, sterility testing

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24

material

mixed cellulose esters (MCE)
plain filter
white filter

sterility

non-sterile

feature

autoclavable
hydrophobic

packaging

box of 10 envelopes containing 10 pak

border W

6 mm , hydrophobic edge

filter diam.

47 mm

pore size

0.45 μm pore size

input

sample(s) (environmental)
sample type bottled water
sample type soft drink
sample type food(s)
sample type cider
sample type beer
sample type cosmetics
sample(s) (environmental)
sample type sport drink
sample type beverage(s)
sample type wine

application(s)

clinical
cosmetics
environmental
food and beverages
industrial qc

shipped in

ambient

General description

Filter Code: HAEP
Hydrophobic edge membranes are standard MF-Type filters with edges. The non-wetting edge prevents drug intrusion under holder sealing rim

Application

Filling machine operations, sterility testing

Packaging

Autoclave pack: 10 resealable envelopes containing 10 filters and 10 pads, requires autoclaving

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service