Skip to Content
Merck
All Photos(1)

Documents

FCMAB188F

Sigma-Aldrich

Milli-Mark® Anti-CD24-FITC Antibody, clone SN3

clone SN3, Milli-Mark®, from mouse

Synonym(s):

Signal transducer CD24, Small cell lung carcinoma cluster 4 antigen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.44

biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

SN3, monoclonal

species reactivity

human

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

Related Categories

General description

The CD24 antigen, which has a molecular weight of 35-45 kDa, is a glycosyl phosphatidylinositol (GPI)-anchored surface molecule. It is a member of a family that also includes HSA and CD52. Cross-linking of CD24 can induce increased intracellular Ca++ in mature B lymphocytes (Turni, 2002).
CD24 is expressed at multiple stages of B cell development, beginning with the bone marrow CD34+ pro-B-cell compartment and continuing through mature surface IgM+/IgD+ B cells. Plasma cells express very low to negligible levels of CD24 (Le Bien, 1995). CD24 is not expressed on the cell surface of normal T cells or monocytes, but is expressed on mature granulocytes (Le Bien, 1995; Fukukawa, 1986). The antigen is also present on a variety of epithelial types (Turni, 2002).

Specificity

Antibody recognizes Human CD24 antigen.

Immunogen

Isolated membrane fraction of the pre-B leukemic cell line NALM-1.

Application

Detect CD24 using this Milli-Mark Anti-CD24-FITC Antibody, clone SN3 validated for use in FC.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Quality

Evaluated by flow cytometry using human PBMCs

Target description

35-45 kDa (Turni, 2002)

Physical form

Protein A purified
Purified Mouse monoclonal IgG1 conjugated to FITC in 0.05M Tris-HCl buffer with 1% BSA, Sodium Azide at 0.097% at pH 7.2.

Storage and Stability

Maintained refrigerated at 2-8°C from date of receipt. Protected from light in undiluted aliquots for 6 months from date of receipt.

Analysis Note

Control
PBMCs

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li Wang et al.
Experimental and therapeutic medicine, 14(5), 4773-4780 (2017-12-05)
Gemcitabine is the first-line chemotherapeutic agent for advanced adenocarcinoma of the pancreas, despite the high risk of chemoresistance as a major disadvantage. In the past few years, significant advances have been made in the field of pancreatic cancer stem-like cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service