Key Documents
EZSPUMP01
Millipore
Liquid Pump
for use with EZ-Stream® Vacuum Pump
Select a Size
1 EACH
7.420,00 kr.
7.420,00 kr.
Please contact Customer Service for Availability
Request more information
Select a Size
Change View
1 EACH
7.420,00 kr.
About This Item
UNSPSC Code:
41116500
NACRES:
NB.79
7.420,00 kr.
Please contact Customer Service for Availability
Request more information
feature
repair kit
manufacturer/tradename
EZ-Stream®
application(s)
bioburden testing
food and beverages
industrial qc
pharmaceutical
water monitoring
compatibility
for use with EZ-Stream® Vacuum Pump
General description
Liquid Pump is a spare part for EZ-Stream® Vacuum Pump, it is used to replace internal liquid pump in case of malfunction or filtration issue to keep optimal filtration performance.
The design of the system allows fast and easy replacement of this component.
The EZ-Stream® Vacuum Pump is ideal to be used both on work bench and laminar flow hood, where its small footprint saves laboratory space.
The kit includes:
  • 1 Liquid Pump.
Application
Liquid Pump is a spare part used for EZ-Stream® Vacuum Pump. The EZ-Stream® Vacuum Pump is used to provide hassle free liquid discard during filtration process.
Legal Information
EZ-STREAM is a registered trademark of Merck KGaA, Darmstadt, Germany