Skip to Content
Merck
All Photos(1)

Key Documents

EZBRAIN40

Millipore

Human Amyloid β40 Brain ELISA

Sign Into View Organizational & Contract Pricing

Select a Size

96 EA
7.430,00 kr.

7.430,00 kr.


Estimated to ship on18 March 2025Details


Request a Bulk Order

Select a Size

Change View
96 EA
7.430,00 kr.

About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

7.430,00 kr.


Estimated to ship on18 March 2025Details


Request a Bulk Order

grade

for protein analysis

Quality Level

description

analytes available: Amyloid β40
16-500 pg/mL (standard curve range)

species reactivity

human

sensitivity measuring range

6.0 pg/mL

technique(s)

ELISA: suitable

detection method

colorimetric

General description

The lowest level of Amyloid β1-40 standard used in this assay is 6.0 pg/mL (50 βL sample size).

Specificity

The Amyloid β40 ELISA uses monoclonal anti-Aβ antibodies with high selectivity for human Aβ. The capture antibody recognizes the C-terminal end of Amyloid β1-40, which causes a high selectivity for Aβ 40. The cross-reactivity of the used antibodies to other Amyloid peptides was tested by ELISA and BIACORE and shows no significant cross-reactivity to Aβ1-38, Aβ1-39, Aβ1-42, Aβ1-43 and Aβ1-44.

Application

This assay requires 50 µl of sample and is an overnight assay.

Storage and Stability

Components in the kit can be stored up to 2 weeks at 2-8°C

Disclaimer

For research use only. Not for use in diagnostic procedures.

Pictograms

CorrosionExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Met. Corr. 1 - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service