Skip to Content
Merck
All Photos(1)

Key Documents

ET-BAGRACK-KIT

Millipore

NovaSeptum® GO Fixture for TC Bag Rack

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
1.417,00 kr.

1.417,00 kr.


Estimated to ship on26 June 2025Details



Select a Size

Change View
1 PKG
1.417,00 kr.

About This Item

UNSPSC Code:
41104105

1.417,00 kr.


Estimated to ship on26 June 2025Details


product line

NovaSeptum® GO

Quality Level

feature

autoclavable

parameter

-80-150 °C temp. range (-112-302 °F)
6 bar max. pressure (87 psi) at ±25 °C

application(s)

pathogen testing
pyrogen testing
sterile sampling
sterility testing

storage temp.

room temp

General description

Device configuration: Sterile Sampling accessory
NovaSeptum® GO Fixture is designed to make compatible TC Bag Rack with NovaSeptum® GO

Application

Sterility testing, pyrogenic testing, endotoxin testing, chemical analysis, pH analysis, and fermentation applications

Packaging

ET-BAGRACK-KIT (1 unit per box)

Components

  • Stainless steel in compliance with A4

Legal Information

NOVASEPTUM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service