Skip to Content
Merck
All Photos(1)

Key Documents

ENSTAG

Millipore

NovaSeptum® GO First Actuation Locking Tag

Sign Into View Organizational & Contract Pricing

Select a Size

100 PKG
1.363,00 kr.

1.363,00 kr.


Estimated to ship on19 May 2025Details



Select a Size

Change View
100 PKG
1.363,00 kr.

About This Item

UNSPSC Code:
41104105

1.363,00 kr.


Estimated to ship on19 May 2025Details


product line

NovaSeptum® GO

feature

autoclavable

parameter

0-134 °C temp. range (32-273 °F)

application(s)

pathogen testing
pyrogen testing
sterile sampling
sterility testing

storage temp.

room temp

General description

Device configuration: Sterile Sampling accessory
NovaSeptum® GO First actuation tag is used to demonstrate the absence of teampering of the sampling solution prior to the first intended actuation.

Application

Sterility testing, pyrogenic testing, endotoxin testing, chemical analysis, pH analysis, and fermentation applications

Packaging

ENSTAG (100 unit per box)

Components

  • Locking tag: Polyamid/stainless steel

Legal Information

NOVASEPTUM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service