ECM610
PAI Activity Assay
The PAI Activity Assay Kit provides a quick, efficient & sensitive system for evaluation of inhibition of uPA activity by PAI & for screening of uPA inhibitors.
Select a Size
4.830,00 kr.
Select a Size
About This Item
4.830,00 kr.
Recommended Products
Quality Level
species reactivity
human
manufacturer/tradename
Chemicon®
technique(s)
activity assay: suitable
NCBI accession no.
UniProt accession no.
shipped in
dry ice
Gene Information
human ... PLAU(5328)
General description
The CHEMICON PAI Activity Assay Kit utilizes a chromogenic substrate, which is cleaved by active uPA, detectable by its Optical Density at 405nm (OD405). Addition of PAI sample blocks the cleavage of substrate by uPA.
The CHEMICON PAI Assay Kit is ideal for measurement of PAI inhibitory activity toward uPA in purified preparations and cell culture, as well as in serum where pathological conditions such as sepsis exist. The assay is also useful for screening inhibitors of uPA.
Each CHEMICON PAI Activity Assay Kit contains sufficient reagents for the evaluation of 96 samples, including uPA standard from human urine and E.Coli-expressed recombinant protein corresponding to amino acids 24 to 427 of human PAI-1 (mature form) as positive control. PAI-1 positive control contains a mixture of the active and latent forms, with >45% of active form. Duplicate or triplicate samples are suggested.
The CHEMICON PAI Activity Assay Kit is intended for research use only; not for diagnostic or therapeutic applications.
Test Principle:
The CHEMICON PAI Activity Assay Kit utilizes a chromogenic substrate, which is cleaved by active uPA, detectable by its Optical Density at 405nm (OD405). Addition of PAI sample blocks the cleavage of substrate by uPA.
Application:
The CHEMICON PAI Assay Kit is ideal for measurement of PAI inhibitory activity toward uPA in purified preparations and cell culture, as well as in serum where pathological conditions such as sepsis exist. The assay is also useful for screening inhibitors of uPA.
Each CHEMICON PAI Activity Assay Kit contains sufficient reagents for the evaluation of 96 samples, including uPA standard from human urine and E. coli expressed recombinant protein corresponding to amino acids 24 to 427 of human PAI-1 (mature form) as positive control. PAI-1 positive control contains a mixture of the active and latent forms, with >45% of active form. Duplicate or triplicate samples are suggested.
The CHEMICON PAI Activity Assay Kit is intended for research use only; not for diagnostic or therapeutic applications.
Application
Cell Structure
Packaging
Components
Chromogenic Substrate: (Part No. 90545) One 5 mg bottle of Tripeptide with pNA group.
Assay Buffer, 10X: (Part No. 90091) One 5 mL bottle.
PAI-1 Positive Control: (Part No. 90093) One 400 μL vial containing 20 μg of purified PAI-1 with 0.1% BSA.
Storage and Stability
Legal Information
Disclaimer
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Resp. Sens. 1
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service