E221
NovaSeptum® GO Autoclavable Bag Sampling System, Vented
sterile; β-irradiated
Synonym(s):
NovaSeptum® GO Autoclavable Bag, Vented
About This Item
Recommended Products
material
316 stainless steel cannula
polyester body
polypropylene bag
silicone septum (platinum-cured)
silicone tubing
Quality Level
Agency
according to ISO 11137 (sterilization)
according to ISO 146441
meets USP 88 Biological Reactivity (all component materials)
sterility
sterile; β-irradiated
product line
NovaSeptum® GO
feature
autoclavable
parameter
-20-125 °C temp. range (-4-257 °F)
0.30 bar max. pressure (4.35 psi)
impurities
<2.15 EU/device bacterial endotoxins (LAL test)
fitting
male Luer-Lok® outlet (with female sterile vent filter)
application(s)
cell therapy
gene therapy
life science and biopharma
mAb
ophthalmics
parenterals
pharma/biopharma processes
sterile sampling
viral therapy
storage temp.
room temp
Looking for similar products? Visit Product Comparison Guide
General description
Application
Autoclaved
Packaging
E221-00216 (100 ml bags, 40 units per box)
E221-C0243 (250 ml bags, 40 units per box)
E221-C0244 (1000 ml bags, 40 units per box)
Components
- Septum: Platinum-cured silicone
- Body: Polyester
- Cannula: ASTM® 316 L Stainless steel
- Fluid contact layer (bag): Polypropylene film
- Tubing: Silicone
- Outlet Tubing: Male Luer-Lok® with female sterile vent filter
Preparation Note
Aqueous extraction contains < 2.15 EU per device as determined using the Limulus Amebocyte Lysate (LAL) test.
Legal Information
Not finding the right product?
Try our Product Selector Tool.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service