Key Documents
CR0003L106
Millipore
Bellco BBC3 CC System style 3L motor adapter
Select a Size
1 EA
inquire
This item is not able to be purchased as a guest user. For further information, please contact us at +45 35 15 80 78or
.
Select a Size
Change View
1 EA
inquire
About This Item
UNSPSC Code:
23151806
eCl@ss:
32031690
This item is not able to be purchased as a guest user. For further information, please contact us at +45 35 15 80 78or
.
Quality Level
General description
Bellco BBC3 CC System style 3L motor adapter for use with Mobius CellReady 3L Bioreactor