Skip to Content
Merck
All Photos(1)

Key Documents

BX1780

Supelco

Butyl Alcohol

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Synonym(s):

Butyl Alcohol, 1-Butanol

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
788,00 kr.
4 L
2.370,00 kr.

788,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 ML
788,00 kr.
4 L
2.370,00 kr.

About This Item

Linear Formula:
CH3(CH2)2CH2OH
CAS Number:
MDL number:
UNSPSC Code:
12352104
EC Index Number:
200-751-6
grade:
reagent
bp:
116-118 °C/1013 hPa
vapor pressure:
6.7 hPa ( 20 °C)

788,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

grade

reagent

Quality Level

Agency

meets USP/NF

vapor pressure

6.7 hPa ( 20 °C)

Assay

≥99.4% (GC)

form

liquid

dilution

(for analytical testing)

impurities

≤0.0008 meq/g Titrable acid
≤0.01% Carbonyl compounds (as butyraldehyde)
≤0.1% Water (H2O)
≤0.2% Di-n-Butyl ether [(C4 H9)2 O]

evapn. residue

≤0.005%

color

APHA: ≤10

pH

7 (20 °C, 70 g/L in H2O)

bp

116-118 °C/1013 hPa

mp

-89 °C

transition temp

flash point 34 °C

solubility

66 g/L

density

0.81 g/cm3 at 20 °C

shipped in

ambient

storage temp.

room temp

SMILES string

OCCCC

InChI

1S/C4H10O/c1-2-3-4-5/h5H,2-4H2,1H3

InChI key

LRHPLDYGYMQRHN-UHFFFAOYSA-N

Application

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Analysis Note

Assay (GC): 99.4% min
Carbonyl compounds (as butyraldehyde): 0.01% max
Color (APHA): 10 max
Di-n-Butyl ether [(C4 H9)2 O]: 0.2% max
Evaporation residue: 0.005% max
Form: Clear liquid
Titrable acid: 0.0008meq/g max
Water (H2O): 0.1% max

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

95.0 °F - Pensky-Martens closed cup

Flash Point(C)

35 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service