Skip to Content
Merck
All Photos(1)

Key Documents

BM-12075

Millipore

Auto2D® PAGE Chip 7.5%

precast gel, for second-dimension separations on the Auto2D® 2-D Electrophoresis Device.

Synonym(s):

2D Electrophoresis, 2DE, PAGE Chip, PAGE gel, SDS-PAGE, precast gel

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
2.270,00 kr.

2.270,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
2.270,00 kr.

About This Item

UNSPSC Code:
41100000
NACRES:
NC.01

2.270,00 kr.


Please contact Customer Service for Availability

General description

PAGE Chip is a Pre-cast gel for SDS-PAGE (2nd dimension). PAGE Chip 7.5% is used for second-dimension separations on the Auto2D® 2-D Electrophoresis Device. This covers 30kDa or higher molecular weight range with focused separation on 70kDa and higher molecular weight.

Application

  • Differential proteomic analysis
  • Post-translational modification
  • Protein Isoforms
  • Western Blotting

Legal Information

Auto2D is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Auto2D® automates difficult 2D electrophoresis methods for protein localization with high reproducibility in under two hours.

Protocols

The Auto2D® Automated 2-D Gel Electrophoresis Device video and detailed protocol provide reliable methods to prepare your samples and operate the Auto2D® Device.

Related Content

Auto2D® 2-D Gel Electrophoresis Device frequently asked questions for 2-D PAGE and additional 2DE protein separation workflow needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service