Skip to Content
Merck
All Photos(1)

Key Documents

BI0344

Sigma-Aldrich

Capping Reagent B ACN/Acetic Anhydride/Lutidine (50/20/30)

Biosynthesis Novabiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116006
Product BI0344 is not currently sold in your country. Contact Technical Service

Quality Level

product line

Novabiochem®

form

liquid

reaction suitability

reaction type: Acetylations
reagent type: derivatization reagent

color

yellow

transition temp

flash point 5 °C

shipped in

ambient

storage temp.

room temp

Application

BI0344-4000 is packaged in 38/430 glass bottles
BI0344-P4000 is packaged in poly coated glass bottles
DNA Reagents Label Change Explanation
Biosynthesis Novabiochem®

Analysis Note

Assay (2,6-lutidine)29.0-31.0% v/vAssay (acetic anhydride)19.0-21.0% v/vAssay (acetonitrile)49.0-51.0% v/vColorYellowFormLiquidInfrared Spectrummatches STDWater (H2O)0.05% max

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

41.0 °F - closed cup

Flash Point(C)

5 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service